Learn More
1-Bromo-3-methylbutane, 99%, Thermo Scientific Chemicals
£16.80 - £67.90
Chemical Identifiers
CAS | 107-82-4 |
---|---|
Molecular Formula | C5H11Br |
Molecular Weight (g/mol) | 151.05 |
MDL Number | MFCD00000253 |
InChI Key | YXZFFTJAHVMMLF-UHFFFAOYSA-N |
Synonym | isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa |
PubChem CID | 7891 |
IUPAC Name | 1-bromo-3-methylbutane |
SMILES | CC(C)CCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10540552
|
Thermo Scientific Acros
403050050 |
5 g | Glass bottle |
£16.80
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10607751
|
Thermo Scientific Acros
403051000 |
100 g | Glass bottle |
£17.90
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10249450
|
Thermo Scientific Acros
403055000 |
500 g | Glass bottle |
£67.90
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
107-82-4 | |
151.05 | |
YXZFFTJAHVMMLF-UHFFFAOYSA-N | |
7891 | |
CC(C)CCBr |
C5H11Br | |
MFCD00000253 | |
isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa | |
1-bromo-3-methylbutane |
Specifications
0.1% max. (as HBr) | |
98.5 | |
-112°C | |
1.2000g/mL | |
32°C | |
98.5% min. (GC) | |
(CH3)2CHCH2CH2Br | |
MFCD00000253 | |
15, 5159 | |
isoamyl bromide, isopentyl bromide, butane, 1-bromo-3-methyl, 3-methylbutyl bromide, 4-bromo-2-methylbutane, isobutylmethyl bromide, isopentylbromide, 1-bromo-3-methyl-butane, iso-pentyl bromide, unii-czm50594qa | |
YXZFFTJAHVMMLF-UHFFFAOYSA-N | |
1-bromo-3-methylbutane | |
7891 | |
99% | |
1-Bromo-3-methylbutane, 99% |
107-82-4 | |
100.0 | |
Colorless | |
120°C to 121°C | |
Authentic | |
C5H11Br | |
1.4399 to 1.4419 | |
01, 136 | |
1.2 | |
Solubility in water: 0.2g/L in water (16°C). Other solubilities: miscible with alcohol,ether,benzene and,carbon tetrachloride | |
CC(C)CCBr | |
151.05 | |
151.05 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
IF ON SKIN: Wash with plenty of soap a
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-522-9
missing translation for 'rtecsNumber' : EJ6230000
missing translation for 'tsca' : TSCA
RUO – Research Use Only