missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromo-4-butoxybenzene, 97%, Thermo Scientific™
Click to view available options
Quantity:
5 g
10 g
50 g
Specifications
Specifications
CAS Min % | 96.5 |
CAS Max % | 100.0 |
Assay Percent Range | 97% |
Linear Formula | BrC6H4O(CH2)3CH3 |
Quantity | 5 g |
Synonym | p-bromophenyl butyl ether, benzene,1-bromo-4-butoxy, p-butoxybromobenzene, benzene, 1-bromo-4-butoxy, 4-bromo-1-butoxybenzene, 4-butoxybromobenzene, 4-n-butoxyphenyl bromide, 1-butoxy-4-bromobenzene, 4-bromophenyl butyl ether |
Formula Weight | 229.12 |
Percent Purity | 97% |
Chemical Name or Material | 1-Bromo-4-butoxybenzene |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'