missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-4-(difluoromethyl)benzene, 97%, Thermo Scientific™

Product Code. 10822421 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 10822421

Brand: Thermo Scientific Acros 432550010

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 51776-71-7
Molecular Formula C7H5BrF2
Molecular Weight (g/mol) 207.02
MDL Number MFCD06657964
InChI Key HUSPSWKWFREKSS-UHFFFAOYSA-N
Synonym 1-bromo-4-difluoromethyl benzene, 1-bromo-4-difluoromethyl-benzene, benzene, 1-bromo-4-difluoromethyl, 4-bromo-alpha,alpha-difluorotoluene, 4-bromobenzal fluoride, pubchem23296, acmc-1aukw, 1-bromo-4-difluoro-methyl-benzene
PubChem CID 2761183
IUPAC Name 1-bromo-4-(difluoromethyl)benzene
SMILES FC(F)C1=CC=C(Br)C=C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 74.0°C to 76.0°C (12.0 Torr)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 1 g
Formula Weight 207.02
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromo-4-(difluoromethyl)benzene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Bromo-4-(difluoromethyl)benzene
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H302-Harmful if swallowed.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.