missing translation for 'onlineSavingsMsg'
Learn More

1-Bromo-5-fluoropentane, 99%, Thermo Scientific Chemicals

Product Code. 15402179
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as ?-opioid receptor imaging agents.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromo-5-fluoropentane was used to prepare 18F-labeled (anilido)piperidine derivatives as μ-opioid receptor imaging agents.

Solubility
Soluble in methanol (in methanol). Slightly soluble in water.

Notes
Store in cool, dry place in tightly closed container. With adequate ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 407-97-6
Molecular Formula C5H10BrF
Molecular Weight (g/mol) 169.037
MDL Number MFCD01709395
InChI Key GMYIZICPHREVDH-UHFFFAOYSA-N
Synonym pentane, 1-bromo-5-fluoro, 5-fluoroamyl bromide, 5-fluoropentyl bromide, fluoropentyl bromide, 5-bromopentyl fluoride, acmc-20a2uo, 1-bromo-5-fluoro-pentane, pentane,1-bromo-5-fluoro, 4-01-00-00313 beilstein handbook reference, ksc493m6b
PubChem CID 120236
IUPAC Name 1-bromo-5-fluoropentane
SMILES C(CCF)CCBr

Specifications

Color Colorless
Density 1.360 g/mL
Boiling Point 162°C
Linear Formula F(CH2)5Br
Refractive Index 1.4406
Quantity 5 g
Beilstein 1697258
Solubility Information Soluble in methanol (in methanol). Slightly soluble in water.
Formula Weight 169.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-Bromo-5-fluoropentane
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'