missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Chemical Identifiers
CAS | 768-90-1 |
---|---|
Molecular Formula | C10H15Br |
Molecular Weight (g/mol) | 215.13 |
MDL Number | MFCD00074721 |
InChI Key | VQHPRVYDKRESCL-UHFFFAOYSA-N |
Synonym | 1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane |
PubChem CID | 79106 |
IUPAC Name | 1-bromoadamantane |
SMILES | C1C2CC3CC1CC(C2)(C3)Br |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10001570
|
Thermo Scientific Acros
149030250 |
25 g | Glass bottle |
£34.47
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10756191
|
Thermo Scientific Acros
149031000 |
100 g | Glass bottle |
£80.60
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
768-90-1 | |
215.13 | |
VQHPRVYDKRESCL-UHFFFAOYSA-N | |
79106 | |
C1C2CC3CC1CC(C2)(C3)Br |
C10H15Br | |
MFCD00074721 | |
1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
1-bromoadamantane |
Specifications
768-90-1 | |
100.0 | |
White to Yellow | |
98.5% min. (GC) | |
MFCD00074721 | |
Solubility in water: insoluble. Other solubilities: soluble in diethyl ether,chloroform,benzene,,methanol and ethanol | |
C1C2CC3CC1CC(C2)(C3)Br | |
215.13 | |
215.13 | |
Powder |
98.5 | |
117.0°C to 120.0°C | |
Authentic | |
C10H15Br | |
1-adamantyl bromide, adamantyl bromide, bromoadamantane, 1-bromo-adamantane, adamantane, 1-bromo, 1-bromotricyclo 3.3.1.13,7 decane, tricyclo 3.3.1.13,7 decane, 1-bromo, tricyclo 3.3.1.1 3,7-decane, 1-bromo, labotest-bb lt00239617, 3r,5s,7s-1-bromoadamantane | |
VQHPRVYDKRESCL-UHFFFAOYSA-N | |
1-bromoadamantane | |
79106 | |
99% | |
1-Bromoadamantane, 99% |
Safety and Handling
MOISTURE SENSITIVE
missing translation for 'einecsNumber' : 212-200-7
missing translation for 'tsca' : TSCA
Product Identifier
- 1-Bromoadamantane
RUO – Research Use Only