missing translation for 'onlineSavingsMsg'
Learn More

1-Bromobenzyl methyl ether, 98%, Thermo Scientific Chemicals

Product Code. 10698984
Click to view available options
Quantity:
1 g
5 g
25 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52711-30-5
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.06
MDL Number MFCD03790887
InChI Key QFAZLCRHLRJNAW-UHFFFAOYSA-N
Synonym 1-bromo-2-methoxymethyl benzene, 2-bromobenzyl methyl ether, acmc-20ag76, methyl 2-bromobenzyl ether, 1-methoxymethyl-2-bromobenzene, 2-bromobenzyl alcohol, methyl ether, benzene,1-bromo-2-methoxymethyl
PubChem CID 12927162
IUPAC Name 1-bromo-2-(methoxymethyl)benzene
SMILES COCC1=CC=CC=C1Br

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Linear Formula BrC6H4CH2OCH3
Refractive Index 1.5470 to 1.5490 (20°C, 589nm)
Quantity 1 g
Formula Weight 201.06
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Bromobenzyl methyl ether
Show More Show Less
compliance-icons
Product Identifier
  • 1-Bromobenzyl methyl ether
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'