missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromohexadecane, 97%, Thermo Scientific Chemicals
£47.20 - £234.00
Chemical Identifiers
CAS | 112-82-3 |
---|---|
Molecular Formula | C16H33Br |
Molecular Weight (g/mol) | 305.34 |
MDL Number | MFCD00000230 |
InChI Key | HNTGIJLWHDPAFN-UHFFFAOYSA-N |
Synonym | hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam |
PubChem CID | 8213 |
IUPAC Name | 1-bromohexadecane |
SMILES | CCCCCCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10104510
|
Thermo Scientific Acros
215432500 |
250 mL | Glass bottle |
£47.20
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10194500
|
Thermo Scientific Acros
215430010 |
1 L | Glass bottle |
£104.00
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11984491
|
Thermo Scientific Acros
215430025 |
2.5 L | Glass bottle |
£234.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-82-3 | |
305.34 | |
HNTGIJLWHDPAFN-UHFFFAOYSA-N | |
8213 | |
CCCCCCCCCCCCCCCCBr |
C16H33Br | |
MFCD00000230 | |
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam | |
1-bromohexadecane |
Specifications
112-82-3 | |
Amber to Black-Brown | |
336.0°C | |
Authentic | |
C16H33Br | |
1.4609 to 1.4629 | |
01,172 | |
hexadecyl bromide, cetyl bromide, n-hexadecyl bromide, hexadecane, 1-bromo, 1-hexadecyl bromide, n-hexadecyl-1-bromide, bromohexadecane, unii-76gji7gvam, hexadecane, bromo, 76gji7gvam | |
HNTGIJLWHDPAFN-UHFFFAOYSA-N | |
1-bromohexadecane | |
8213 | |
97% | |
1-Bromohexadecane |
17.3°C | |
0.9990g/mL | |
177°C | |
96% min. (GC) | |
CH3(CH2)15Br | |
MFCD00000230 | |
0.999 | |
Solubility in water: insoluble. Other solubilities: soluble in most organic solvents | |
CCCCCCCCCCCCCCCCBr | |
305.34 | |
305.33 | |
Liquid After Melting |
Safety and Handling
missing translation for 'einecsNumber' : 204-008-7
RUO – Research Use Only