Learn More
Chemical Identifiers
CAS | 111-25-1 |
---|---|
Molecular Formula | C6H13Br |
Molecular Weight (g/mol) | 165.07 |
MDL Number | MFCD00000271 |
InChI Key | MNDIARAMWBIKFW-UHFFFAOYSA-N |
Synonym | hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga |
PubChem CID | 8101 |
IUPAC Name | 1-bromohexane |
SMILES | CCCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10429590
|
Thermo Scientific Acros
107045000 |
500 mL | Glass bottle |
£32.66
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10529990
|
Thermo Scientific Acros
107040025 |
2.5 L | Glass bottle |
£117.59
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11904491
|
Thermo Scientific Acros
107040100 |
10 L | Plastic drum |
£357.78
10 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
111-25-1 | |
165.07 | |
MNDIARAMWBIKFW-UHFFFAOYSA-N | |
8101 | |
CCCCCCBr |
C6H13Br | |
MFCD00000271 | |
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga | |
1-bromohexane |
Specifications
0.1% max. (as HBr) | |
99.0 | |
-85°C | |
154°C to 158°C | |
Authentic | |
C6H13Br | |
1.4465 to 1.4485 | |
01,144 | |
hexyl bromide, n-hexyl bromide, bromohexane, 1-hexyl bromide, hexane, 1-bromo, hexane, bromo, unii-wva0fax7ga, 1-bromo-hexane, pentane, bromomethyl, wva0fax7ga | |
MNDIARAMWBIKFW-UHFFFAOYSA-N | |
1-bromohexane | |
8101 | |
99+% | |
1-Bromohexane |
111-25-1 | |
100.0 | |
1.1700g/mL | |
57°C | |
0.5% max. Hexanol (GC) | |
CH3(CH2)5Br | |
MFCD00000271 | |
1.17 | |
Solubility in water: insoluble | |
CCCCCCBr | |
165.07 | |
165.07 | |
Liquid |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
Flammable liquid and vapor.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 203-850-2
- 1-Bromohexane
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only