missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-(Bromomethyl)-3-phenoxybenzene, 97%, Thermo Scientific™
£52.95 - £302.00
Chemical Identifiers
CAS | 51632-16-7 |
---|---|
Molecular Formula | C13H11BrO |
Molecular Weight (g/mol) | 263.134 |
InChI Key | UJUNUASMYSTBSK-UHFFFAOYSA-N |
Synonym | 1-bromomethyl-3-phenoxybenzene, m-phenoxybenzyl bromide, 3-phenoxybenzyl bromide, benzene, 1-bromomethyl-3-phenoxy, alpha-bromo-3-phenoxytoluene, 3-phenoxybenzylbromide, unii-gd31x56z15, 3-phenyloxybenzylbromide, m-bromomethyl phenyl phenyl ether, m-phenoxybenzylbromide |
PubChem CID | 94544 |
IUPAC Name | 1-(bromomethyl)-3-phenoxybenzene |
SMILES | C1=CC=C(C=C1)OC2=CC=CC(=C2)CBr |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10507273
|
Thermo Scientific Maybridge
MO07329CB |
250 mg |
£52.95
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10021962
|
Thermo Scientific Maybridge
MO07329DA |
1 g |
£113.00
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10768645
|
Thermo Scientific Maybridge
MO07329DE |
5 g |
£302.00
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Chemical Identifiers
51632-16-7 | |
263.134 | |
1-bromomethyl-3-phenoxybenzene, m-phenoxybenzyl bromide, 3-phenoxybenzyl bromide, benzene, 1-bromomethyl-3-phenoxy, alpha-bromo-3-phenoxytoluene, 3-phenoxybenzylbromide, unii-gd31x56z15, 3-phenyloxybenzylbromide, m-bromomethyl phenyl phenyl ether, m-phenoxybenzylbromide | |
1-(bromomethyl)-3-phenoxybenzene |
C13H11BrO | |
UJUNUASMYSTBSK-UHFFFAOYSA-N | |
94544 | |
C1=CC=C(C=C1)OC2=CC=CC(=C2)CBr |
Specifications
51632-16-7 | |
C13H11BrO | |
UJUNUASMYSTBSK-UHFFFAOYSA-N | |
1-(bromomethyl)-3-phenoxybenzene | |
94544 | |
97% |
97% | |
1-bromomethyl-3-phenoxybenzene, m-phenoxybenzyl bromide, 3-phenoxybenzyl bromide, benzene, 1-bromomethyl-3-phenoxy, alpha-bromo-3-phenoxytoluene, 3-phenoxybenzylbromide, unii-gd31x56z15, 3-phenyloxybenzylbromide, m-bromomethyl phenyl phenyl ether, m-phenoxybenzylbromide | |
C1=CC=C(C=C1)OC2=CC=CC(=C2)CBr | |
263.134 | |
263.13 | |
1-(Bromomethyl)-3-phenoxybenzene |