missing translation for 'onlineSavingsMsg'
Learn More

1-(Bromomethyl)-4-phenoxybenzene, ≥97%, Thermo Scientific™

£82.80 - £200.00

Chemical Identifiers

CAS 36881-42-2
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.134
MDL Number MFCD04038693
InChI Key CPIGBCFBFZSCQI-UHFFFAOYSA-N
PubChem CID 1514251
IUPAC Name 1-(bromomethyl)-4-phenoxybenzene
SMILES C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
10642515
View Documents
Thermo Scientific Maybridge
CC53708CB
250 mg
£82.80
250mg
Please to purchase this item. Need a web account? Register with us today!
10154323
View Documents
Thermo Scientific Maybridge
CC53708DA
1 g
£200.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

36881-42-2
263.134
CPIGBCFBFZSCQI-UHFFFAOYSA-N
1-(bromomethyl)-4-phenoxybenzene
C13H11BrO
MFCD04038693
1514251
C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr

Specifications

36881-42-2
100.0
C13H11BrO
1-bromomethyl-4-phenoxybenzene, 4-phenoxybenzyl bromide, 1-bromomethyl-4-phenoxy-benzene, benzene,1-bromomethyl-4-phenoxy, 4-phenyloxybenzylbromide, 4-bromomethyl-1-phenoxybenzene, benzene, 1-bromomethyl-4-phenoxy
C1=CC=C(C=C1)OC2=CC=C(C=C2)CBr
263.134
263.13
1-(Bromomethyl)-4-phenoxybenzene
96.5
97%
MFCD04038693
CPIGBCFBFZSCQI-UHFFFAOYSA-N
1-(bromomethyl)-4-phenoxybenzene
1514251
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Harmful if swallowed.
Causes serious eye damage.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.