missing translation for 'onlineSavingsMsg'
Learn More
1-Bromonaphthalene, 96%, Thermo Scientific Chemicals Product Code.: 10234420

1-Bromonaphthalene, 96%, Thermo Scientific Chemicals

Product Code. 10234420
100 mL, Glass bottle
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
Packaging:
Glass bottle
This item is not returnable. View return policy

Product Code. 10234420

Brand: Thermo Scientific Acros 107151000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-11-9, 580-13-2
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthaleneShow MoreShow Less
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

CAS Min % 95.0
CAS Max % 100.0
Color Brown-Yellow to Yellow
Infrared Spectrum Authentic
Assay Percent Range 96%
Packaging Glass bottle
Molecular Formula C10H7Br
Linear Formula C10H7Br
Refractive Index 1.6560 to 1.6590 (20°C, 589nm)
Quantity 100 mL
Beilstein 05, 547
Merck Index 15, 1436
Formula Weight 207.07
Vapor Pressure 14.8hPa at 20°C
Percent Purity ≥95%
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene, 96%
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0469104 Certificate TypeCertificate of Analysis Date31/01/2025 Catalog Number107150025,107151000,107155000
Lot NumberA0462233 Certificate TypeCertificate of Analysis Date23/04/2024 Catalog Number107150050,107150025,107151000,107155000
Lot NumberA0461208 Certificate TypeCertificate of Analysis Date18/03/2024 Catalog Number107150050,107150025,107151000,107155000
Lot NumberA0460987 Certificate TypeCertificate of Analysis Date08/03/2024 Catalog Number107150050,107150025,107151000,107155000
Lot NumberA0460386 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog Number107150050,107150025,107151000,107155000
compliance-icons compliance-icons
Product Identifier
  • 1-Bromonaphthalene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Bromonaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromonaphthalene, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.