missing translation for 'onlineSavingsMsg'
Learn More
1-Bromonaphthalene, 97%, Thermo Scientific Chemicals Product Code.: 11408096

1-Bromonaphthalene, 97%, Thermo Scientific Chemicals

Product Code. 11408096
250 g
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy

Product Code. 11408096

Brand: Thermo Scientific Alfa Aesar A14122.30

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Bromonaphthalene is used in organic synthesis and refractometry of fats. It is used in the preparation of N-aryl imidazoles and diaryl ethers. It is used to determine the refractive index of crystals and the water content of alcohols. It is utilized in the preparation of glutathione peroxidase-like antioxidant activity of diaryl diselenides. Further, it serves as a solvent for the exfoliation and dispersion of hexabenzocoronene. In addition to this, it is used as a refrigerant and a solvent molecular weight substance.

Solubility
Miscible with chloroform, ethanol, ether, benzene and ethyl acetate. Slightly miscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-11-9
Molecular Formula C10H7Br
Molecular Weight (g/mol) 207.07
MDL Number MFCD00003868
InChI Key DLKQHBOKULLWDQ-UHFFFAOYSA-N
Synonym naphthalene, 1-bromo, 1-naphthyl bromide, alpha-bromonaphthalene, bromonaphthalene, alpha-naphthyl bromide, .alpha.-bromonaphthalene, 1-bromo-naphthalene, .alpha.-naphthyl bromide, naphthalene, bromo, bromonapthalene
PubChem CID 7001
IUPAC Name 1-bromonaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Br

Specifications

Melting Point -2°C to 0°C
Density 1.485
Boiling Point 279°C to 281°C
Flash Point >110°C (230°F)
Assay Percent Range 97%
Refractive Index 1.657
Quantity 250 g
Beilstein 1906414
Merck Index 14,1425
Solubility Information Miscible with chloroform,ethanol,ether,benzene and ethyl acetate. Slightly miscible with water.
Formula Weight 207.08
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-Bromonaphthalene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberA0462233 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA14122.0B,A14122.18,A14122.30
Lot NumberA0460987 Certificate TypeCertificate of Analysis Date16/04/2024 Catalog NumberA14122.0B,A14122.18,A14122.30
Lot NumberA0458232 Certificate TypeCertificate of Analysis Date07/02/2024 Catalog NumberA14122.0B,A14122.18,A14122.30
Lot NumberA0456260 Certificate TypeCertificate of Analysis Date31/01/2024 Catalog NumberA14122.0B,A14122.18,A14122.30
Lot NumberA0456474 Certificate TypeCertificate of Analysis Date25/10/2023 Catalog NumberA14122.0B,A14122.18,A14122.30
compliance-icons compliance-icons
Product Identifier
  • 1-Bromonaphthalene
Signal Word
  • Warning
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 1
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H319-Causes serious eye irritation.
  • H410-Very toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Bromonaphthalene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Bromonaphthalene, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.