missing translation for 'onlineSavingsMsg'
Learn More

1-Bromooctane, 99%, Thermo Scientific Chemicals

£58.50 - £211.00

Chemical Identifiers

CAS 111-83-1
Molecular Formula C8H17Br
Molecular Weight (g/mol) 193.13
MDL Number MFCD00000276
InChI Key VMKOFRJSULQZRM-UHFFFAOYSA-N
Synonym n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
PubChem CID 8140
IUPAC Name 1-bromooctane
SMILES CCCCCCCCBr
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10071910
View Documents Promotion Details
Thermo Scientific Acros
166815000
500 mL Glass bottle
£58.50
500mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15740819
View Documents Promotion Details
Thermo Scientific Acros
166810025
2.5 L Glass bottle
£211.00
2.50 L
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

111-83-1
193.13
VMKOFRJSULQZRM-UHFFFAOYSA-N
8140
CCCCCCCCBr
C8H17Br
MFCD00000276
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
1-bromooctane

Specifications

111-83-1
1.1100g/mL
78°C
99%
CH3(CH2)7Br
MFCD00000276
15,6853
n-octyl bromide, octyl bromide, 1-octylbromide, octane, 1-bromo, bromooctane, 1-bromo-octane, 1-bromo octane, 1-bromo-n-octane, unii-q382hpc6lc, octane, bromo
VMKOFRJSULQZRM-UHFFFAOYSA-N
1-bromooctane
8140
99%
1-Bromooctane
-55.0°C
201.0°C
Authentic
C8H17Br
1.4508 to 1.4528
01,16
1.11
Solubility in water: insoluble. Other solubilities: miscible with alcohol and ether
CCCCCCCCBr
193.13
193.13
Liquid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Avoid release to the environment.
Collect spillage.
Dispose of contents/container to an approved waste disposal plant.

GHS Signal Word: Warning

missing translation for 'einecsNumber' : 203-912-9

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only