Learn More
1-Bromopropane, 99%, Thermo Scientific Chemicals
£11.50 - £141.00
Chemical Identifiers
CAS | 106-94-5 |
---|---|
Molecular Formula | C3H7Br |
MDL Number | MFCD00000254 |
InChI Key | CYNYIHKIEHGYOZ-UHFFFAOYSA-N |
Synonym | n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide |
PubChem CID | 7840 |
ChEBI | CHEBI:47105 |
IUPAC Name | 1-bromopropane |
SMILES | CCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10724791
|
Thermo Scientific Acros
107310050 |
5 mL | Glass bottle | N/A | |||||
10305980
|
Thermo Scientific Acros
107312500 |
250 mL | Glass bottle |
£20.70
250mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10428250
|
Thermo Scientific Acros
107315000 |
500 mL | Glass bottle |
£34.90
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10617252
|
Thermo Scientific Acros
107310010 |
1 L | Glass bottle |
£60.40
1 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10175403
|
Thermo Scientific Acros
107310025 |
2.5 L | Glass bottle |
£141.00
2.50 L |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
106-94-5 | |
MFCD00000254 | |
n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide | |
CHEBI:47105 | |
CCCBr |
C3H7Br | |
CYNYIHKIEHGYOZ-UHFFFAOYSA-N | |
7840 | |
1-bromopropane |
Specifications
106-94-5 | |
100.0 | |
Authentic | |
C3H7Br | |
1.4320 to 1.4360 (20°C, 589nm) | |
n-propyl bromide, propyl bromide, propane, 1-bromo, propane, bromo, bromopropane, 1-bromo-propane, 1-propyl bromide, ccris 30, unii-y9746dne68, n-propylbromide | |
CCCBr | |
122.993 | |
CHEBI:47105 | |
≥98.5% | |
1-Bromopropane |
98.5 | |
Colorless to Yellow | |
99% | |
CH3CH2CH2Br | |
MFCD00000254 | |
CYNYIHKIEHGYOZ-UHFFFAOYSA-N | |
1-bromopropane | |
7840 | |
122.99 | |
Liquid |
Safety and Handling
GHS H Statement:
Causes serious eye irritation.
May cause drowsiness or dizziness.
Causes skin irritation.
May cause damage to organs through prolonged or repeated exposure.
May damage fertility. May damage the unborn child.
May cause respiratory irritation.
Highly flamma
GHS P Statement:
Obtain special instructions before use.
IF exposed or concerned: Get medical advice/attention.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification
Danger
Recommended Storage : Product darkens during storage
- 1-Bromopropane
Signal Word
- Danger
Hazard Category
- Flammable liquid Category 2
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Carcinogenicity Category 2
- Reproductive toxicity Category 1B
- Specific target organ toxicity Category 3
- Specific target organ toxicity after repeated exposure Category 2
- LONG-TERM AQUATIC HAZARD Chronic 3
Hazard Statement
- H225-Highly flammable liquid and vapour.
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H336-May cause drowsiness or dizziness.
- H351-Suspected of causing cancer.
- H360FD-May damage fertility. May damage the unborn child.
- H373-May cause damage to organs.
- H412-Harmful to aquatic life with long lasting effects.
- H420-Harms public health and the environment by destroying ozone in the upper atmosphere.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
- P502-Refer to manufacturer/supplier for information on recovery/recycling.
RUO – Research Use Only