missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Bromotetradecane, 98%, Thermo Scientific Chemicals
£37.29 - £649.35
Chemical Identifiers
CAS | 112-71-0 |
---|---|
Molecular Formula | C14H29Br |
Molecular Weight (g/mol) | 277.28 |
MDL Number | MFCD00000228 |
InChI Key | KOFZTCSTGIWCQG-UHFFFAOYSA-N |
Synonym | myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo |
PubChem CID | 8208 |
IUPAC Name | 1-bromotetradecane |
SMILES | CCCCCCCCCCCCCCBr |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10377700
|
Thermo Scientific Acros
187381000 |
100 g | Glass bottle |
£37.29
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10489680
|
Thermo Scientific Acros
187385000 |
500 g | Glass bottle |
£159.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13488418
|
Thermo Scientific Acros
187380025 |
2.5 kg | Glass bottle | N/A | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
112-71-0 | |
277.28 | |
KOFZTCSTGIWCQG-UHFFFAOYSA-N | |
8208 | |
CCCCCCCCCCCCCCBr |
C14H29Br | |
MFCD00000228 | |
myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo | |
1-bromotetradecane |
Specifications
112-71-0 | |
100.0 | |
Yellow | |
175.0°C to 178.0°C (20.0 mmHg) | |
Authentic | |
C14H29Br | |
1.4590 to 1.4610 | |
01,II,136 | |
myristyl bromide, tetradecyl bromide, tetradecane, 1-bromo, n-tetradecyl bromide, 1-tetradecyl bromide, bromotetradecane, n-tetradecyl-1-bromide, 1-bromo tetradecane, 1-bromo-n-tetradecane, tetradecane, bromo | |
KOFZTCSTGIWCQG-UHFFFAOYSA-N | |
1-bromotetradecane | |
8208 | |
98% | |
1-Bromotetradecane, 98% |
97.5 | |
5.0°C to 6.0°C | |
0.9300g/mL | |
149°C | |
97.5% min. (GC) | |
CH3(CH2)13Br | |
MFCD00000228 | |
0.93 | |
Solubility in water: insoluble | |
CCCCCCCCCCCCCCBr | |
277.28 | |
277.28 | |
Liquid |
Safety and Handling
missing translation for 'einecsNumber' : 203-999-3
missing translation for 'tsca' : TSCA
Product Identifier
- 1-Bromotetradecane
RUO – Research Use Only