missing translation for 'onlineSavingsMsg'
Learn More
1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals Product Code.: 11423847

1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals

Product Code. 11423847
5 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11423847

Brand: Thermo Scientific Alfa Aesar A18295.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2386-60-9
Molecular Formula C4H9ClO2S
Molecular Weight (g/mol) 156.624
MDL Number MFCD00007464
InChI Key WEDIIKBPDQQQJU-UHFFFAOYSA-N
Synonym 1-butanesulfonyl chloride, butylsulfonyl chloride, butanesulfonyl chloride, 1-butanesulfonylchloride, 1-butylsulfonyl chloride, n-butanesulphonyl chloride, butanesulfonylchloride, butane-1-sulphonyl chloride, n-butylsulfonyl chloride, n-butanesulfonyl chloride
PubChem CID 75430
IUPAC Name butane-1-sulfonyl chloride
SMILES CCCCS(=O)(=O)Cl

Specifications

Melting Point -29°C
Density 1.21
Boiling Point 80°C to 81°C (9 mmHg)
Flash Point 79°C (174°F)
Linear Formula CH3(CH2)3SO2Cl
Refractive Index 1.456
Quantity 5 g
UN Number UN3265
Beilstein 1748742
Sensitivity Moisture sensitive
Formula Weight 156.63
Percent Purity 98%
Chemical Name or Material 1-Butanesulfonyl chloride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249138 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog NumberA18295.06,A18295.22,A18295.14
Lot Number10245087 Certificate TypeCertificate of Analysis Date19/12/2022 Catalog NumberA18295.06,A18295.22,A18295.14
Lot NumberI8775A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18295.06,A18295.22,A18295.14
Lot Number10050785 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18295.06,A18295.22,A18295.14
Lot Number10175186 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberA18295.06,A18295.22,A18295.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanesulfonyl chloride, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.