missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, 99.9%, Honeywell™

Product Code. 15615800
100 mL, Glass Bottle
Click to view available options
Quantity:
100 mL
1 L
2.5 L
20 L
4 x 2.5 L
6 x 1 L
Packaging:
Aluminum bottle
Glass Bottle
Stainless Steel Can
This item is not returnable. View return policy
This item is not returnable. View return policy

≥99.9%

  • We are your supplier of high-quality lab reagents and chemicals for analytical and chemistry laboratories:
    Solvents – ACS specific use (HPLC), ACS general use, solvents for histology, and reagent grade for chemical synthesis and other industrial applications.
    Inorganics – Chemical synthesis and inorganic chemistry including essential acids and bases, salts, metals and elements, and reagents for chemical reactions.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -78°C
Appearance Colorless liquid
Density 0.81g/mL at 25°C
Boiling Point 101°C to 105°C
Quantity 100 mL
Explosion Limit 11.2%
Ignition Point 649°C
Packaging Glass Bottle
Linear Formula CH3(CH2)3OH
UN Number UN1120
Molecular Weight (g/mol) 74.12
Formula Weight 74.12g/mol
Percent Purity 99%
Chemical Name or Material 74.12g/mol
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, 99.9%, Honeywell™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.