missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, ACS, 99.4+%

Product Code. 11337388
Click to view available options
Quantity:
500 mL
1 L
4 L
4 x 1 L
4 x 4 L
Unit Size:
1 L
16 L
4 L
500mL
This item is not returnable. View return policy

Product Code. 11337388

Brand: Thermo Scientific Alfa Aesar 031068.M1

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Color (APHA) 10, Evaporation residue 0.005%, Titratable acid 0.0008meq/g, Carbonyl compounds (as butyraldehyde) 0.01%, Butyl ether 0.2%, H2O 0.1% | CAS: 71-36-3 | C4H10O | 74.123 g/mol

1-Butanol is used as solvent for many organic reactions in pharmaceutical and chemical industry, as a solvent in coating industries and also used as a diluent/reactant in the manufacture of urea-formaldehyde and melamine-formaldehyde resins. It is used in the production of drugs, alkaloids, additive in floor cleaners, consumer products and stain removers. It is used as mobile phase in paper and thin layer chromatography. It is also used as an alternative for gasoline and diesel fuels.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Color Colorless
Appearance Colorless liquid
Density 0.81
Boiling Point 117°C to 118°C
Quantity 4 x 1 L
Flash Point 35°C (95°F)
Odor Alcool-like
Physical Form Liquid
Refractive Index 1.399
Assay Percent Range >99.4%
Vapor Pressure 5.6 hPa (4mm Hg) at 20°C
Beilstein 969148
Solubility Information Miscible with alcohols,ketones,aldehydes,ethers,aliphatic and aromatic hydrocarbons. Slightly miscible with water.
Formula Weight 74.12
Grade ACS
Concentration or Composition (by Analyte or Components) 1-Butanol
Chemical Name or Material 1-Butanol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.