missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™

Product Code. 15624620
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C
Density 0.81g/mL at 25°C
Boiling Point 116°C to 118°C
Quantity 4 x 2.5 L
Refractive Index n20/D 1.399
Linear Formula CH3(CH2)3OH
MDL Number MFCD00002964
Beilstein 969148
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
Molecular Weight (g/mol) 74.123
Formula Weight 74.12g/mol
Percent Purity ≥99.7%
Grade CHROMASOLV™ Plus
Chemical Name or Material 1-Butanol, for HPLC
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'