missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%, Honeywell Riedel-de Haën™

Product Code. 15634620
Click to view available options
Quantity:
100 mL
1 L
2 L
2.5 L
4 x 2.5 L
6 x 1 L

CHROMASOLV™ Plus, for HPLC,≥99.7%

  • For HPLC, spectrophotometry, and environmental analysis
  • Tested for HPLC-suitability via gradient analysis and UV-VIS spectroscopy
  • Low UV impurity profile
  • Low impurity gradient LC baseline
  • Very low levels of non-volatile impurities
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO

Specifications

Melting Point -90°C (literature)
Absorbance max. 0.10 at 240 nm, max. 0.10 at 235 nm, max. 0.50 at 225 nm, max. 0.70 at 220 nm, max. 1.0 at 210 nm, max. 0.01 at 400 nm, max. 0.01 280 nm, max. 0.04 at 260 nm
CAS Max % 100.0000%
Density 0.81 g/mL at 25 °C(lit.)
Boiling Point 119°C
Quantity 6 x 1 L
Flash Point 35°C
Packaging Glass Bottle
Linear Formula C6H5CH3
MDL Number MFCD00008512
UN Number UN1120
Beilstein 969148
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropane
Molecular Weight (g/mol) 74.123
Formula Weight 74.12
Percent Purity ≥99.7%
Grade HPLC
Chemical Name or Material 1-Butanol, CHROMASOLV™ Plus, for HPLC, ≥99.7%
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'