missing translation for 'onlineSavingsMsg'
Learn More

1-Butanol, Honeywell

£62.14 - £1027.30

Chemical Identifiers

CAS 71-36-3
Molecular Formula C4H10O
Molecular Weight (g/mol) 74.123
MDL Number MFCD00002964
InChI Key LRHPLDYGYMQRHN-UHFFFAOYSA-N
Synonym 1-butanol, butanol, n-butanol, butyl alcohol, n-butyl alcohol, 1-hydroxybutane, propylcarbinol, butyl hydroxide, propylmethanol, methylolpropaneShow MoreShow Less
PubChem CID 263
ChEBI CHEBI:28885
IUPAC Name butan-1-ol
SMILES CCCCO
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15653880
SDS View Documents
Honeywell Riedel-de Haën
33065-1L
1 L
£62.14
1 L
Please to purchase this item. Need a web account? Register with us today!
15663880
SDS View Documents Promotion Details
Honeywell Riedel-de Haën
33065-2.5L
2.5 L
£147.06
2.50 L
Please to purchase this item. Need a web account? Register with us today!
15673880
SDS View Documents
Honeywell Riedel-de Haën
33065-30KG
30 kg
£1027.30
30kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Butanol, Honeywell

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.