Learn More
1-Butyl-3-methylimidazolium chloride, 98+%, Thermo Scientific Chemicals
£20.70 - £439.00
Chemical Identifiers
CAS | 79917-90-1 |
---|---|
Molecular Formula | C8H15ClN2 |
Molecular Weight (g/mol) | 174.67 |
MDL Number | MFCD03095425 |
InChI Key | FHDQNOXQSTVAIC-UHFFFAOYSA-M |
Synonym | 1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461 |
PubChem CID | 2734161 |
IUPAC Name | 1-butyl-3-methylimidazol-3-ium;chloride |
SMILES | [Cl-].CCCCN1C=C[N+](C)=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10671784
|
Thermo Scientific Acros
354090050 |
5 g |
£20.70
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10490242
|
Thermo Scientific Acros
354090250 |
25 g |
£53.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10463825
|
Thermo Scientific Acros
354091000 |
100 g |
£142.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
10619853
|
Thermo Scientific Acros
354095000 |
500 g |
£439.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Ionic liquidChemical Identifiers
79917-90-1 | |
174.67 | |
FHDQNOXQSTVAIC-UHFFFAOYSA-M | |
2734161 | |
[Cl-].CCCCN1C=C[N+](C)=C1 |
C8H15ClN2 | |
MFCD03095425 | |
1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461 | |
1-butyl-3-methylimidazol-3-ium;chloride |
Specifications
79917-90-1 | |
100.0 | |
Cream to Yellow | |
98% min. (HPLC) | |
C8H15ClN2 | |
15, 1584 | |
Solubility in water: soluble | |
[Cl-].CCCCN1C=C[N+](C)=C1 | |
174.67 | |
170.1 mPa.s (80°C) | |
98+% | |
1-Butyl-3-methylimidazolium chloride |
98.0 | |
73.0°C | |
Authentic | |
Glass bottle | |
MFCD03095425 | |
1-butyl-3-methylimidazolium chloride, 3-butyl-1-methyl-1h-imidazol-3-ium chloride, 1-n-butyl-3-methylimidazolium chloride, bmimcl, bmim cl, c4mim chloride, dsstox_cid_11461, dsstox_rid_78878, dsstox_gsid_31461 | |
FHDQNOXQSTVAIC-UHFFFAOYSA-M | |
1-butyl-3-methylimidazol-3-ium;chloride | |
2734161 | |
174.67 | |
Mass |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Toxic if swallowed.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep a
GHS Signal Word: Danger
missing translation for 'tsca' : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.
- 1-Butyl-3-methylimidazolium chloride
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 3
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H335-May cause respiratory irritation.
- H301-Toxic if swallowed.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P332+P313-If skin irritation occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
RUO – Research Use Only