Learn More
1-Butyl-3-methylimidazolium tetrafluoroborate, 98+%, Thermo Scientific Chemicals
£33.27 - £232.16
Chemical Identifiers
CAS | 174501-65-6 |
---|---|
Molecular Formula | C8H15BF4N2 |
Molecular Weight (g/mol) | 226.03 |
MDL Number | MFCD03095449 |
InChI Key | LSBXQLQATZTAPE-UHFFFAOYSA-N |
Synonym | 1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate |
PubChem CID | 2734178 |
IUPAC Name | 1-butyl-3-methylimidazol-3-ium;tetrafluoroborate |
SMILES | F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10551282
|
Thermo Scientific Acros
354210050 |
5 g | Glass bottle |
£33.27
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10549643
|
Thermo Scientific Acros
354210250 |
25 g | Glass bottle |
£82.41
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10330904
|
Thermo Scientific Acros
354211000 |
100 g | Glass bottle |
£232.16
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Ionic liquidChemical Identifiers
174501-65-6 | |
226.03 | |
LSBXQLQATZTAPE-UHFFFAOYSA-N | |
2734178 | |
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1 |
C8H15BF4N2 | |
MFCD03095449 | |
1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate | |
1-butyl-3-methylimidazol-3-ium;tetrafluoroborate |
Specifications
174501-65-6 | |
100.0 | |
Orange-Yellow | |
Authentic | |
C8H15BF4N2 | |
15, 1584 | |
1-butyl-3-methylimidazolium tetrafluoroborate, 1-n-butyl-3-methylimidazolium tetrafluoroborate, 3-butyl-1-methyl-1h-imidazol-3-ium tetrafluoroborate, unii-t2tvz2306t, bmimbf4, butylmethylimidazolium tetrafluoroborate, 1-butyl-3-methylimidazolium terafluoroborate, 1-methyl-3-butylimidazolium tetrafluoroborate, c4mim tetrafluoroborate, 1-butyl-3-methyl-1h-imidazol-3-ium tetrafluoroborate | |
F[B-](F)(F)F.CCCCN1C=C[N+](C)=C1 | |
226.03 | |
118.3 mPa.s (25°C) | |
98+% | |
1-Butyl-3-methylimidazolium tetrafluoroborate |
98.0 | |
-71.0°C | |
1.2077g/mL | |
98% min. (HPLC) | |
MFCD03095449 | |
1.2077 | |
LSBXQLQATZTAPE-UHFFFAOYSA-N | |
1-butyl-3-methylimidazol-3-ium;tetrafluoroborate | |
2734178 | |
226.02 | |
Viscous Liquid |
Safety and Handling
GHS H Statement
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful to aquatic life with long lasting effects.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical adv
GHS Signal Word: Danger
missing translation for 'tsca' : TSCA
- 1-Butyl-3-methylimidazolium tetrafluoroborate
Signal Word
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only