missing translation for 'onlineSavingsMsg'
Learn More

1-Butylphosphonic acid, 98%

Product Code. 11489116
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11489116

Brand: Thermo Scientific Alfa Aesar A17983.14

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3321-64-0 | C4H11O3P | 138.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3321-64-0
Molecular Formula C4H11O3P
Molecular Weight (g/mol) 138.10
MDL Number MFCD00012295
InChI Key UOKRBSXOBUKDGE-UHFFFAOYSA-N
Synonym n-butylphosphonic acid, 1-butanephosphonic acid, butylphosphonate, 1-butylphosphonic acid, phosphonic acid, butyl, n-butyl phosphonic acid, acmc-1cr41
PubChem CID 76839
ChEBI CHEBI:41384
IUPAC Name butylphosphonic acid
SMILES CCCCP(O)(O)=O

Specifications

Melting Point 105°C to 107°C
Odor Odorless
Quantity 25 g
UN Number UN3261
Beilstein 1749026
Formula Weight 138.1
Percent Purity 98%
Chemical Name or Material 1-Butylphosphonic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Vielen Dank, dass Sie uns helfen, unsere Website zu verbessern. Ihr Feedback wurde übermittelt