missing translation for 'onlineSavingsMsg'
Learn More

1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate, 98+%, Thermo Scientific™

Product Code. 15420956
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 135540-11-3
Molecular Formula C9H16ClF6N2P
Molecular Weight (g/mol) 332.66
MDL Number MFCD00191333
InChI Key NHEGCUSBUWGOQM-UHFFFAOYSA-N
Synonym pyclu, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium hexafluorophosphate, chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro pyrrolidin-1-yl methylene pyrrolidin-1-ium hexafluorophosphate v, 1-chloro-1-pyrrolidinylmethylene pyrrolidinium, pyciu, chlorodipyrrolidinocarbenium hexafluorophosphate, chloro-n,n,n',n'-bis tetramethylene formamidinium hexafluorophosphate, chloro-dipyrrolidinocarbenium hexafluorophosphate, pyclu chlorodipyrrolidinocarbenium hexafluorophosphate, 1-chloro-1-pyrrolidinylmethylene pyrrolidiniumhexafluorophosphate
PubChem CID 2736652
IUPAC Name 1-[chloro(pyrrolidin-1-ium-1-ylidene)methyl]pyrrolidine;hexafluorophosphate
SMILES F[P-](F)(F)(F)(F)F.ClC(N1CCCC1)=[N+]1CCCC1

Specifications

Melting Point 153°C to 156°C
Quantity 1 g
Beilstein 7898575
Sensitivity Light and moisture sensitive
Formula Weight 332.65
Percent Purity ≥98%
Chemical Name or Material 1-(Chloro-1-pyrrolidinylmethylene)pyrrolidinium hexafluorophosphate
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'