missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether, 97%, Thermo Scientific Chemicals

Product Code. 11328386
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether is a useful solvent for dispersing fluorinated materials.

For Research and Development use only. Not to be used in clinical studies

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26675-46-7
Molecular Formula C3H2ClF5O
Molecular Weight (g/mol) 184.49
MDL Number MFCD00066609
InChI Key PIWKPBJCKXDKJR-UHFFFAOYNA-N
Synonym isoflurane, forane, 1-chloro-2,2,2-trifluoroethyl difluoromethyl ether, forene, aerrane, isoflurano, isofluranum, isoflo, compound 469, 2-chloro-2-difluoromethoxy-1,1,1-trifluoroethane
PubChem CID 3763
ChEBI CHEBI:6015
IUPAC Name 2-chloro-2-(difluoromethoxy)-1,1,1-trifluoroethane
SMILES FC(F)OC(Cl)C(F)(F)F

Specifications

Color Colorless
Density 1.496g/mL
Boiling Point 48°C to 49°C
Odor Ether-like
Refractive Index <1.3
Quantity 25 g
Beilstein 1852087
Merck Index 14,5175
Formula Weight 184.49
Percent Purity 97%
Chemical Name or Material 1-Chloro-2,2,2-trifluoroethyl difluoromethyl ether
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'