missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-2-methylpropyl chloroformate, 98%, Thermo Scientific Chemicals

Product Code. 10781345
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Coupling reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 92600-11-8
Molecular Formula C5H8Cl2O2
Molecular Weight (g/mol) 171.02
MDL Number MFCD00012387
InChI Key BOGLZTPABVHWDD-BYPYZUCNSA-N
Synonym 1-chloro-2-methylpropyl chloroformate, acmc-20aow3, 1-chloro-2-methylpropylchloroformate, 1-chloro-2-methylpropyl carbonochloridate, chloro 1-chloro-2-methylpropoxy methanone, carbonochloridic acid,1-chloro-2-methylpropyl ester
PubChem CID 2733326
SMILES CC(C)[C@@H](Cl)OC(Cl)=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless
Density 1.1700g/mL
Flash Point 53°C
Infrared Spectrum Authentic
Assay Percent Range 98%
Packaging Glass bottle
Linear Formula ClCO2CH(Cl)CH(CH3)2
Refractive Index 1.4310 to 1.433
Quantity 5 g
Specific Gravity 1.17
IUPAC Name (1-chloro-2-methylpropyl) carbonochloridate
Formula Weight 171.02
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-chloro-2-methylpropyl chloroformate
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'