missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3,4-dinitrobenzene, tech. 90%

Product Code. p-7026660
Click to view available options
Quantity:
50 g
100 g
250 g
Unit Size:
100g
250g
50g
This item is not returnable. View return policy

Product Code. 11479807

Brand: Thermo Scientific Alfa Aesar B22773.18

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 610-40-2 | C6H3ClN2O4 | 202.55 g/mol

1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3,4-dinitrobenzene is used as pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 610-40-2
Molecular Formula C6H3ClN2O4
Molecular Weight (g/mol) 202.55
MDL Number MFCD00007212
InChI Key QVQSOXMXXFZAKU-UHFFFAOYSA-N
Synonym 1-chloro-3,4-dinitrobenzene, 3,4-dinitrochlorobenzene, benzene, 4-chloro-1,2-dinitro, unii-z29b1f274e, 1,2-dinitro-4-chlorobenzene, 1,2-dino2 4-cl benzene, 4-chlordinitrobenzol, acmc-209mog, 4-chloro-1,2-dinitrobenzen, ghl.pd_mitscher_leg0.928
PubChem CID 33097
IUPAC Name 4-chloro-1,2-dinitrobenzene
SMILES C1=CC(=C(C=C1Cl)[N+](=O)[O-])[N+](=O)[O-]

Specifications

Melting Point ∼20°C to 25°C
Boiling Point 14°C to 16°C (4 mmHg)
Flash Point >110°C (230°F)
Quantity 50 g
UN Number UN1577
Beilstein 521564
Formula Weight 202.55
Percent Purity 90%
Grade Technical
Chemical Name or Material 1-Chloro-3,4-dinitrobenzene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.