missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-3-iodopropane, 98%, stab. with copper, Thermo Scientific Chemicals

Product Code. 11475573
Click to view available options
Quantity:
25 g
100 g
500 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-3-iodopropane has been used in the synthesis of N-[4-[5-(2,4-diamino-6-oxo-1,6-dihydropyrimidin-5-yl)-2-(2,2,2-trifluoroacetyl)pentyl]benzoyl]-L-glutamic acid, an inhibitor of glycinamide ribonucleotide transformylase (GAR Tfase) and aminoimidazole carboxamide ribonucleotide transformylase (AICAR Tfase), interesting proton sponge type molecule quino[7,8-h]quinoline.

Solubility
Difficult to mix.

Notes
Light Sensitive. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6940-76-7
Molecular Formula C3H6ClI
Molecular Weight (g/mol) 204.435
MDL Number MFCD00001096
InChI Key SFOYQZYQTQDRIY-UHFFFAOYSA-N
Synonym propane, 1-chloro-3-iodo, 3-chloropropyl iodide, trimethylene chloroiodide, 1-chloro-3-iodo-propane, 3-chloroiodopropane, 3-chloro-iodopropane, 1-chloro-3iodopropane, 1-choro-3-iodopropane, 3-chloro-l-iodopropane, 1-iodo-3-chloropropane
PubChem CID 81363
IUPAC Name 1-chloro-3-iodopropane
SMILES C(CCl)CI

Specifications

Density 1.904
Boiling Point 170°C to 172°C
Flash Point >110°C (230°F)
Refractive Index 1.5463
Quantity 100 g
Beilstein 1731115
Sensitivity Light sensitive
Solubility Information Difficult to mix.
Formula Weight 204.44
Percent Purity 98%
Chemical Name or Material 1-Chloro-3-iodopropane, stabilized with copper
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.