missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloro-3-methyl-2-butene, 95%, stab. with potassium carbonate, Thermo Scientific Chemicals
Click to view available options
Quantity:
10 g
50 g
Description
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.
Solubility
Miscible with chloroform, acetone, diethyl ether and alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
1-Chloro-3-methyl-2-butene is used in the synthesis of geraniol. It is also used as an alkylating agent and as a reagent in hyperforin. Further, it is used as an antibiotic to inhibit growth of tumor cells.
Solubility
Miscible with chloroform, acetone, diethyl ether and alcohol. Immiscible with water.
Notes
Incompatible with strong oxidizing agents and strong acids. Store in a cool place.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Density | 0.928 |
Boiling Point | 107°C to 109°C |
Flash Point | 13°C (56°F) |
Linear Formula | (CH3)2C=CHCH2Cl |
Refractive Index | 1.449 |
Quantity | 10 g |
UN Number | UN1993 |
Solubility Information | Miscible with chloroform,acetone,diethyl ether and alcohol. Immiscible with water. |
Formula Weight | 104.57 |
Percent Purity | 95% |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'