missing translation for 'onlineSavingsMsg'
Learn More

1-Chloro-5-phenylpentane, 99%, Thermo Scientific™

Product Code. 11382666
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 15733-63-8
Molecular Formula C11H15Cl
Molecular Weight (g/mol) 182.69
MDL Number MFCD00037118
InChI Key UXJLBVYYDZDPBV-UHFFFAOYSA-N
Synonym 1-chloro-5-phenylpentane, 5-chloropentyl benzene, benzene, 5-chloropentyl, 5-phenyl-1-pentyl chloride, 5-phenylpentyl chloride, acmc-20aovs, 5-chloro-1-phenylpentane, 5-chloropentyl benzene #
PubChem CID 519173
IUPAC Name 5-chloropentylbenzene
SMILES ClCCCCCC1=CC=CC=C1

Specifications

Density 1.014
Boiling Point 76°C to 79°C (0.65 mmHg)
Refractive Index 1.514
Quantity 1 g
Beilstein 2355978
Formula Weight 182.7
Percent Purity 99%
Chemical Name or Material 1-Chloro-5-phenylpentane
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'