missing translation for 'onlineSavingsMsg'
Learn More
1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals Product Code.: 11381847

1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals

Product Code. 11381847
10 g
Click to view available options
Quantity:
10 g
50 g
This item is not returnable. View return policy

Product Code. 11381847

Brand: Thermo Scientific Alfa Aesar L05198.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloro-6-iodohexane is used as :Pharmaceutical intermediates.

Solubility
Not miscible in water.

Notes
Store at -20°C. Light Sensitive. Store away from oxidizing agents. Incompatible materials are Strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 34683-73-3
Molecular Formula C6H12ClI
Molecular Weight (g/mol) 246.516
MDL Number MFCD00039417
InChI Key QTJHNJCILMMRIQ-UHFFFAOYSA-N
Synonym hexane, 1-chloro-6-iodo, hexamethylene chloroiodide, 6-chlorohexyl iodide, acmc-20aovr, 6-chloro-1-iodohexane, 1-iodo-6-chloro hexane, 1-chloro-6-iodohexane, 1-chloro-6-iodohexane, stab. with copper chip, stabilized with copper chip
PubChem CID 118713
IUPAC Name 1-chloro-6-iodohexane
SMILES C(CCCI)CCCl

Specifications

Refractive Index 1.525
Quantity 10 g
Beilstein 1732408
Sensitivity Light sensitive
Solubility Information Not miscible in water.
Formula Weight 246.52
Percent Purity 97%
Chemical Name or Material 1-Chloro-6-iodohexane

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10243429 Certificate TypeCertificate of Analysis Date13/09/2022 Catalog NumberL05198.09,L05198.18
Lot NumberG10R015 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05198.09,L05198.18
Lot Number10232457 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05198.09,L05198.18
Lot NumberI05R008 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL05198.09,L05198.18
Lot Number10192262 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL05198.09,L05198.18

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chloro-6-iodohexane, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.