missing translation for 'onlineSavingsMsg'
Learn More
1-Chlorobutane, 99+%, Thermo Scientific Chemicals Product Code.: 11430403

1-Chlorobutane, 99+%, Thermo Scientific Chemicals

Product Code. 11430403
500 mL
Click to view available options
Quantity:
500 mL
2500 mL
This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

Product Code. 11430403

Brand: Thermo Scientific Alfa Aesar A11494.AP

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as an alkylating reagent in organic and organometallic compound synthesis. It is also employed as an anthelmintic in veterinary medicine.

Solubility
Miscible with alcohol and ether. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutaneShow MoreShow Less
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point °C to 123°C
Density 0.886
Boiling Point 77°C to 78°C
Flash Point -12°C (10°F)
Assay Percent Range >99%
Refractive Index 1.4024
Quantity 500 mL
UN Number UN1127
Beilstein 1730909
Merck Index 14,1560
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 92.57
Percent Purity ≥99%
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10212637 Certificate TypeCertificate of Analysis Date30/08/2024 Catalog NumberA11494.0F,A11494.AP
Lot Number10133538 Certificate TypeCertificate of Analysis Date10/01/2023 Catalog NumberA11494.0F,A11494.AP
Lot Number10243678 Certificate TypeCertificate of Analysis Date13/10/2022 Catalog NumberA11494.0F,A11494.AP
Lot Number10190930 Certificate TypeCertificate of Analysis Date07/06/2022 Catalog NumberA11494.0F,A11494.AP
Lot Number10184103 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA11494.0F,A11494.AP
compliance-icons compliance-icons
Product Identifier
  • 1-Chlorobutane
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 3
  • Aspiration toxicity Category 1
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H412-Harmful to aquatic life with long lasting effects.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P331-Do NOT induce vomiting.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
Supplemental information
  • MIXTURE LIST-Contains : 1-Chlorobutane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Chlorobutane, 99+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.