missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorobutane, HPLC Grade, 99.5+%

Product Code. 11323588
Click to view available options
Quantity:
250 mL
1 L
4 L
4 x 1 L
Unit Size:
1 L
250mL
4 L
This item is not returnable. View return policy

Product Code. 11323588

Brand: Thermo Scientific Alfa Aesar 039198.K2

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Maximum level of impurities: Evaporation residue 1ppm, H2O 0.02% | CAS: 109-69-3 | C4H9Cl | 92.57 g/mol

1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as alkylating reagent in organic and organometallic compound synthesis. It is also employed as an antihelmintic in veterinary medicine.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-69-3
Molecular Formula C4H9Cl
Molecular Weight (g/mol) 92.57
MDL Number MFCD00001009
InChI Key VFWCMGCRMGJXDK-UHFFFAOYSA-N
Synonym butyl chloride, n-butyl chloride, butane, 1-chloro, chlorure de butyle, nbc wormer, n-propylcarbinyl chloride, sure shot, butane, chloro, chlorobutane, n-chlorobutane
PubChem CID 8005
IUPAC Name 1-chlorobutane
SMILES CCCCCl

Specifications

Melting Point °C to 123°C
Color Colorless
Density 0.886
Boiling Point 77°C to 78°C
Flash Point -12°C (10°F)
Ignition Point 280°C (536°F)
Assay Percent Range >99.5%
Odor Pungent
Refractive Index 1.4024
Quantity 1 L
Beilstein 1730909
Solubility Information Miscible with alcohol and ether. Immiscible with water.
Formula Weight 92.57
Vapor Pressure 110 hPa (83mm Hg) at 20°C
Grade HPLC
Concentration or Composition (by Analyte or Components) 1-Chlorobutane
Physical Form Liquid
Chemical Name or Material 1-Chlorobutane, Filtered through 0.2μ filters.
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.