missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chlorobutane, ReagentPlus™, 99%, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Description
- ≥95% for general laboratory use.
Specifications
Melting Point | -123°C |
Color | Colorless |
Density | 0.886g/mL at 25°C |
Boiling Point | 78.8°C |
Explosion Limit | 0.101 |
Flash Point | −12°C (10.4°F) |
Ignition Point | 860°F |
Packaging | Glass Bottle |
Linear Formula | CH3(CH2)3Cl |
Refractive Index | n20/D 1.402(lit.) |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'