Learn More
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 96%, Thermo Scientific Chemicals
£29.30 - £276.00
Chemical Identifiers
CAS | 140681-55-6 |
---|---|
Molecular Formula | C7H14B2ClF9N2 |
Molecular Weight (g/mol) | 354.26 |
MDL Number | MFCD00142607 |
InChI Key | TXRPHPUGYLSHCX-UHFFFAOYSA-N |
Synonym | selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate |
PubChem CID | 2724933 |
IUPAC Name | 1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate |
SMILES | F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10561881
|
Thermo Scientific Acros
291490050 |
5 g | Glass bottle |
£29.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10202322
|
Thermo Scientific Acros
291490250 |
25 g | Glass bottle |
£96.50
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11926531
|
Thermo Scientific Acros
291491000 |
100 g | Glass bottle |
£276.00
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
C-X bond formation reagentChemical Identifiers
140681-55-6 | |
354.26 | |
TXRPHPUGYLSHCX-UHFFFAOYSA-N | |
2724933 | |
F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 |
C7H14B2ClF9N2 | |
MFCD00142607 | |
selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate | |
1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate |
Specifications
140681-55-6 | |
White | |
Authentic | |
C7H14B2ClF9N2 | |
15,8562 | |
Solubility in water: soluble. Other solubilities: soluble in hcl,moderately soluble in ch3cn,dmf,slightly soluble in (ch3)2co | |
F[B-](F)(F)F.F[B-](F)(F)F.F[N+]12CC[N+](CCl)(CC1)CC2 | |
354.26 | |
354.26 | |
Powder |
260.0°C | |
>54°C | |
95% min. (Iodometry) | |
MFCD00142607 | |
selectfluor, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane bis tetrafluoroborate, f-teda-bf4, 1-chloromethyl-4-fluoro-1,4-diazabicyclo 2.2.2 octane-1,4-diium tetrafluoroborate, unii-4p1za6r76d, selectfluor fluorinating reagent, f-teda, selectfluor r fluorinating reagent, 1-chloromethyl-4-fluoro-1,4-diazoniabicyclo 2.2.2 octane ditetrafluoroborate | |
TXRPHPUGYLSHCX-UHFFFAOYSA-N | |
1-(chloromethyl)-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane;ditetrafluoroborate | |
2724933 | |
96% | |
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) |
Safety and Handling
GHS H Statement
May cause an allergic skin reaction.
Harmful if swallowed.
Harmful to aquatic life with long lasting effects.
Causes serious eye damage.
GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protecti
GHS Signal Word: Danger
- 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo¢2.2.2!octane bis(tetrafluoroborate)
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 3
- Serious eye damage/eye irritation Category 1
- Respiratory sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H412-Harmful to aquatic life with long lasting effects.
- H302-Harmful if swallowed.
Precautionary Statement
- P273-Avoid release to the environment.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
RUO – Research Use Only