missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate), 98+%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) is used as a reagent for electrophilic fluorination and also acts as a selective oxidizing agent. It is a highly effective and versatile source of electrophilic fluorine and also used in the synthesis of organofluorine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) is used as a reagent for electrophilic fluorination and also acts as a selective oxidizing agent. It is a highly effective and versatile source of electrophilic fluorine and also used in the synthesis of organofluorine.
Solubility
Soluble in water, acetonitrile, alcohol, acetone, dimethylformamide, pyridine and dimethylsulfoxide.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases.
1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) is used as a reagent for electrophilic fluorination and also acts as a selective oxidizing agent. It is a highly effective and versatile source of electrophilic fluorine and also used in the synthesis of organofluorine.
Solubility
Soluble in water, acetonitrile, alcohol, acetone, dimethylformamide, pyridine and dimethylsulfoxide.
Notes
Moisture sensitive. Incompatible with strong oxidizing agents, reducing agents, strong acids and bases.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | ∽250°C (decomposition) |
Quantity | 5 g |
UN Number | UN3088 |
Solubility Information | Soluble in water,acetonitrile,alcohol,acetone,dimethylformamide,pyridine and dimethylsulfoxide. |
Formula Weight | 354.26 |
Percent Purity | ≥98% |
Chemical Name or Material | 1-Chloromethyl-4-fluoro-1,4-diazoniabicyclo[2.2.2]octane bis(tetrafluoroborate) |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'