missing translation for 'onlineSavingsMsg'
Learn More

1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene, Thermo Scientific Chemicals

Product Code. 11427277
Click to view available options
Quantity:
50 g
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chloronaphthalene was used as solvent additive in the fabrication of fullerene based photovoltaic devices. It was used in preparation of polyvinyl chloride based membranes of 5,10,15,20-tetrakis(4-methoxyphenyl)porphyrinatocobalt(II) for arsenite determination. Also used in the synthesis of pharmaceutical compounds such as aryl chlorides.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 90-13-1
Molecular Formula C10H7Cl
Molecular Weight (g/mol) 162.616
MDL Number MFCD00003874
InChI Key JTPNRXUCIXHOKM-UHFFFAOYSA-N
Synonym alpha-chloronaphthalene, naphthalene, 1-chloro, 1-naphthyl chloride, chloronaphthalene, 1-chloro-naphthalene, alpha-naphthyl chloride, 1-chlornaftalen, 1-chloro naphthalene, alpha-chlornaphthalene, .alpha.-chloronaphthalene
PubChem CID 7003
IUPAC Name 1-chloronaphthalene
SMILES C1=CC=C2C(=C1)C=CC=C2Cl

Specifications

Melting Point ∼-8°C
Density 1.194
Boiling Point 259°C
Flash Point 121°C (249°F)
Refractive Index 1.633
Quantity 50 g
Beilstein 970836
Merck Index 14,2149
Solubility Information Insoluble in water.
Formula Weight 162.62
Percent Purity 85%
Grade Technical
Assay remainder 2-chloronaphthalene
Chemical Name or Material 1-Chloronaphthalene
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloronaphthalene, tech. 85%, remainder 2-chloronaphthalene
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P270-Do not eat, drink or smoke when using this product.
  • P271-Use only outdoors or in a well-ventilated area.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P330-Rinse mouth.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P391-Collect spillage.
  • P403+P233-Store in a well-ventilated place. Keep container tightly closed.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : 1-Chloronaphthalene,2-chloronaphthalene

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'