missing translation for 'onlineSavingsMsg'
Learn More

1-Chlorophthalazine, 97%

Product Code. 11446260
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 11446260

Brand: Thermo Scientific Alfa Aesar H27143.09

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 5784-45-2 | C8H5ClN2 | 164.592 g/mol

1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1-Chlorophthalazine, is used as a pharmaceutical intermediate and are used as medical isotopes.

Solubility
Insoluble in water. Soluble in dichloromethane, ethanol and methanol.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away strong oxidizing agents. Water, heat, moisture sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5784-45-2
Molecular Formula C8H5ClN2
Molecular Weight (g/mol) 164.592
MDL Number MFCD00024141
InChI Key UCOVESIAFFGEOR-UHFFFAOYSA-N
Synonym phthalazine, 1-chloro, 1-chloro-phthalazine, chlorophthalazine, ccris 7361, 1-chloro-2,3-benzodiazine, 1chlorophthalazine, 1-chlorophthalazin, acmc-20a2sh, 1-chloro phthalazine, 1-chlorophthalazine 1g
PubChem CID 160793
IUPAC Name 1-chlorophthalazine
SMILES C1=CC=C2C(=C1)C=NN=C2Cl

Specifications

Melting Point 109°C to 112°C
Quantity 10 g
Sensitivity Moisture sensitive
Solubility Information Insoluble in water. Soluble in dichloromethane,ethanol and methanol.
Formula Weight 164.59
Percent Purity 97%
Chemical Name or Material 1-Chlorophthalazine
compliance-icons
Product Identifier
  • 1-Chlorophthalazine
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 1-Chlorophthalazine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.