missing translation for 'onlineSavingsMsg'
Learn More

1-Chloropropane, 99%, Thermo Scientific Chemicals

Product Code. 10244470
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 540-54-5
Molecular Formula C3H7Cl
Molecular Weight (g/mol) 78.54
MDL Number MFCD00000995
InChI Key SNMVRZFUUCLYTO-UHFFFAOYSA-N
Synonym propyl chloride, propane, 1-chloro, n-propyl chloride, chloropropane, unii-tuv7462nwk, 1-chloro-propane, tuv7462nwk, n-propylchloride, propane, chloro, 1-chloropropyl
PubChem CID 10899
IUPAC Name 1-chloropropane
SMILES CCCCl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -123°C
Color Colorless
Density 0.8920g/mL
Boiling Point 46°C to 47°C
Flash Point -18°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3CH2CH2Cl
Refractive Index 1.3870 to 1.389
Quantity 5 mL
Beilstein 01, 104
Merck Index 15, 7960
Specific Gravity 0.892
Solubility Information Solubility in water: 2.7g/L (20°C). Other solubilities: soluble in alcohol,ether and benzene
Formula Weight 78.54
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 1-chloropropane
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • 1-Chloropropane
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Flammable liquid Category 2
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'