missing translation for 'onlineSavingsMsg'
Learn More
1-Cyclohexene-1,2-dicarboxylic anhydride, 97+%, Thermo Scientific Chemicals Product Code.: 15493478

1-Cyclohexene-1,2-dicarboxylic anhydride, 97+%, Thermo Scientific Chemicals

Product Code. 15493478
100 g
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy

Product Code. 15493478

Brand: Thermo Scientific Alfa Aesar H56929.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the synthesis of novel glycyrrhetinic acid derivatives on il-1β-induced prostaglandin e2production in normal human dermal fibroblasts and also in the total synthesis of 6,7—Dihydroligustilide.

Solubility
Reacts with water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2426-02-0
Molecular Formula C8H8O3
Molecular Weight (g/mol) 152.15
MDL Number MFCD00005917
InChI Key HMMBJOWWRLZEMI-UHFFFAOYSA-N
Synonym 3,4,5,6-tetrahydrophthalic anhydride, 4,5,6,7-tetrahydroisobenzofuran-1,3-dione, 1-cyclohexene-1,2-dicarboxylic anhydride, 1,3-isobenzofurandione, 4,5,6,7-tetrahydro, unii-01d3m89a2w, cyclohexene-1,2-dicarboxylic anhydride, 3,4,5,6-tetrahydrophthalic acid anhydride, 1,3,4,5,6,7-hexahydro-2-benzofuran-1,3-dione, 1, 4,5,6,7-tetrahydro, 4,5,6,7-tetrahydro-1,3-isobenzofurandioneShow MoreShow Less
PubChem CID 12475
IUPAC Name 4,5,6,7-tetrahydro-2-benzofuran-1,3-dione
SMILES O=C1OC(=O)C2=C1CCCC2

Specifications

Melting Point 69°C to 73°C
Quantity 100 g
UN Number UN2698
Beilstein 136335
Solubility Information Reacts with water.
Formula Weight 152.15
Percent Purity ≥97%
Chemical Name or Material 1-Cyclohexene-1,2-dicarboxylic anhydride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot Number5007F04W Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56929.14,H56929.22
Lot NumberBF060501A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH56929.14,H56929.22

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
1-Cyclohexene-1,2-dicarboxylic anhydride, 97+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.