Learn More
1-Cyclohexene-1-carboxylic acid, 97%, Thermo Scientific Chemicals
£30.10 - £400.00
Chemical Identifiers
CAS | 636-82-8 |
---|---|
Molecular Formula | C7H10O2 |
Molecular Weight (g/mol) | 126.155 |
MDL Number | MFCD00001545 |
InChI Key | NMEZJSDUZQOPFE-UHFFFAOYSA-N |
Synonym | 1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid |
PubChem CID | 69470 |
ChEBI | CHEBI:29565 |
IUPAC Name | cyclohexene-1-carboxylic acid |
SMILES | C1CCC(=CC1)C(=O)O |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11419622
|
Thermo Scientific Alfa Aesar
A10741.03 |
1 g |
£30.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11429622
|
Thermo Scientific Alfa Aesar
A10741.06 |
5 g |
£77.90
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11439622
|
Thermo Scientific Alfa Aesar
A10741.14 |
25 g |
£400.00
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Cyclohexene-1-carboxylic acid is an intermediate in benzoic acid anaerobic decomposition by methanogenic consortium.
Solubility
Soluble in water (0.7 g/L at 20°C).
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with bases and oxidizing agents.
Chemical Identifiers
636-82-8 | |
126.155 | |
NMEZJSDUZQOPFE-UHFFFAOYSA-N | |
69470 | |
cyclohexene-1-carboxylic acid |
C7H10O2 | |
MFCD00001545 | |
1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid | |
CHEBI:29565 | |
C1CCC(=CC1)C(=O)O |
Specifications
636-82-8 | |
1.1 | |
>110°C (230°F) | |
1.499 | |
UN3261 | |
1-cyclohexene-1-carboxylic acid, 1-cyclohexenecarboxylic acid, cyclohex-1-enecarboxylic acid, cyclohexenecarboxylic acid, cyclohex-1-ene-1-carboxylic acid, 1-cyclohexenecarboxylicacid, cyclohexenyl carboxylic acid, 1-cyclohexene carboxylic acid, acmc-1b731, 3,4,5,6-tetrahydrobenzoic acid | |
NMEZJSDUZQOPFE-UHFFFAOYSA-N | |
cyclohexene-1-carboxylic acid | |
69470 | |
126.16 | |
1-Cyclohexene-1-carboxylic acid |
30°C to 34°C | |
138°C to 140°C (14 mmHg) | |
C7H10O2 | |
MFCD00001545 | |
1905520 | |
Soluble in water (0.7g/L at 20°C). | |
C1CCC(=CC1)C(=O)O | |
126.155 | |
CHEBI:29565 | |
97% |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only