missing translation for 'onlineSavingsMsg'
Learn More

1-Cyclohexyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10181222
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

Chemical Identifiers

CAS 135906-00-2
Molecular Formula C16H17NO3
Molecular Weight (g/mol) 271.316
MDL Number MFCD00277770
InChI Key DTLUKEHVTWNAGE-UHFFFAOYSA-N
Synonym 1-cyclohexyl-4-oxo-1,4-dihydroquinoline-3-carboxylic acid, 1-cyclohexyl-4-oxo-3-quinolinecarboxylic acid, maybridge1_006860, acmc-1c0s9, 1-cyclohexyl-4-oxidanylidene-quinoline-3-carboxylic acid
PubChem CID 2729105
IUPAC Name 1-cyclohexyl-4-oxoquinoline-3-carboxylic acid
SMILES C1CCC(CC1)N2C=C(C(=O)C3=CC=CC=C32)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 271.31
Percent Purity 97%
Chemical Name or Material 1-Cyclohexyl-4-oxo-1, 4-dihydroquinoline-3-carboxylic acid
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'