missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester, 97%, Thermo Scientific™
£119.94 - £439.45
Chemical Identifiers
CAS | 1233526-60-7 |
---|---|
Molecular Formula | C14H23BN2O2 |
Molecular Weight (g/mol) | 262.16 |
InChI Key | OCBNABJVDPSMQY-UHFFFAOYSA-N |
Synonym | 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl |
PubChem CID | 56776730 |
IUPAC Name | 1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
SMILES | B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3 |
Description
Chemical Identifiers
1233526-60-7 | |
262.16 | |
1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole |
C14H23BN2O2 | |
OCBNABJVDPSMQY-UHFFFAOYSA-N | |
56776730 | |
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3 |
Specifications
1233526-60-7 | |
96% min. (GC) | |
C14H23BN2O2 | |
OCBNABJVDPSMQY-UHFFFAOYSA-N | |
1-cyclopentyl-4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyrazole | |
56776730 | |
97% |
38°C to 40°C | |
Glass Bottle | |
1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-1h-pyrazole-4-boronic acid, pinacol ester, 1-cyclopentyl-1h-pyrazole-4-boronic acid pinacol ester, 1-cyclopentyl-1h-pyrazol-4-yl boronic acid pinacol ester, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-tetramethyl-1,3,2-dioxaborolan-2-yl pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl-1h-pyrazol, 1h-pyrazole, 1-cyclopentyl-4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl | |
B1(OC(C(O1)(C)C)(C)C)C2=CN(N=C2)C3CCCC3 | |
262.16 | |
262.16 | |
1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester |
Safety and Handling
GHS Signal Word: Warning
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- 1-Cyclopentyl-1H-pyrazole-4-boronic acid pinacol ester
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H302-Harmful if swallowed.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only