missing translation for 'onlineSavingsMsg'
Learn More

1-(Cyclopropylcarbonyl)piperazine, 97%, Thermo Scientific Chemicals

Product Code. 15421398
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 59878-57-8
Molecular Formula C8H15N2O
Molecular Weight (g/mol) 155.22
MDL Number MFCD06369645
InChI Key KIALFUYSJAAJSU-UHFFFAOYSA-O
Synonym 1-cyclopropylcarbonyl piperazine, cyclopropyl piperazin-1-yl methanone, 1-cyclopropanecarbonylpiperazine, cyclopropyl-piperazin-1-yl-methanone, 1-cyclopropanecarbonyl piperazine, piperazine, 1-cyclopropylcarbonyl, piperazinocyclopropyl ketone, cyclopropyl piperazinyl ketone, 1-cyclopropylcarbonylpiperazine, 4-cyclopropanecarbonyl-piperazine
PubChem CID 2064235
IUPAC Name cyclopropyl(piperazin-1-yl)methanone
SMILES O=C(C1CC1)N1CC[NH2+]CC1

Specifications

Color Brown to Colorless
Density 1.087 g/mL
Boiling Point 120°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.524
Quantity 250 mg
Formula Weight 154.21
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material 1-(Cyclopropylcarbonyl)piperazine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'