missing translation for 'onlineSavingsMsg'
Learn More

(+)-1-Deoxymannojirimycin hydrochloride, Thermo Scientific Chemicals

Product Code. p-7042075
Click to view available options
Quantity:
5 mg
10 mg
25 mg
Unit Size:
10mg
25mg
5mg
This item is not returnable. View return policy

Product Code. 15453889

Brand: Thermo Scientific Alfa Aesar J61277.#0

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective inhibitor of a-mannosidase

Selective inhibitor of a-mannosidase

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 73465-43-7
Molecular Formula C6H14ClNO4
Molecular Weight (g/mol) 199.631
MDL Number MFCD00083611
InChI Key ZJIHMALTJRDNQI-MVNLRXSJSA-N
Synonym DMJ; 1,5-Dideoxy-1,5-imino-D-mannitol hydrochloride
PubChem CID 11390018
IUPAC Name (2R,3R,4R,5R)-2-(hydroxymethyl)piperidine-3,4,5-triol;hydrochloride
SMILES C1C(C(C(C(N1)CO)O)O)O.Cl

Specifications

Type (+)-1-Deoxymannojirimycin Hydrochloride
Physical Form Powder
Color White
Quantity 5 mg
Formula Weight 199.6
Chemical Name or Material (+)-1-Deoxymannojirimycin hydrochloride

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.