missing translation for 'onlineSavingsMsg'
Learn More

(+)-1-Deoxynojirimycin

Product Code. 15444939
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
This item is not returnable. View return policy

Product Code. 15444939

Brand: Thermo Scientific Alfa Aesar J62602.MC

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 19130-96-2 | C6H13NO4 | 163.17 g/mol

An alpha-glucosidase inhibitor. Interferes with normal processing of N-linked glycoproteins.(+)-1-Deoxynojirimycin acts as an inhibitor of alfa-glucosidase I and II and maltase-glucoamylase. It also inhibits mammalian glucosidase, intestinal and lysosmal, beta-glucosidase from sweet almonds, pancreatic alfa-amylase and amyloglucosidase. Further, it serves as a enzyme enhancer for the treatment of Fabry and Pompe disease.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Solubility
Soluble in water, dimethyl sulfoxide and methanol.

Notes
Incompatible with strong acids and strong bases.
TRUSTED_SUSTAINABILITY

Specifications

Type (+)-1-Deoxynojirimycin
Physical Form Powder
CAS 19130-96-2
Melting Point 195°C to 196°C
Color White
Quantity 100 mg
Molecular Formula C6H13NO4
MDL Number MFCD00063474
Merck Index 142905
Synonym 1,5-Dideoxy-1,5-imino-D-glucitol; (2R,3R,4R,5S)-2-(Hydroxymethyl)-3,4,5-piperidinetriol
Solubility Information Soluble in water,dimethyl sulfoxide and methanol.
InChI Key LXBIFEVIBLOUGU-JGWLITMVSA-N
SMILES OC[C@H]1NC[C@H](O)[C@@H](O)[C@@H]1O
IUPAC Name (2R,3R,4R,5S)-2-(hydroxymethyl)piperidine-3,4,5-triol
Molecular Weight (g/mol) 163.17
PubChem CID 29435
ChEBI CHEBI:44369
Formula Weight 163.17
Chemical Name or Material (+)-1-Deoxynojirimycin
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.