Learn More
1-Dimethylamino-2-nitroethylene, 98%, Thermo Scientific Chemicals
£23.10 - £83.20
Chemical Identifiers
CAS | 1190-92-7 |
---|---|
Molecular Formula | C4H8N2O2 |
Molecular Weight (g/mol) | 116.12 |
MDL Number | MFCD00051519 |
InChI Key | JKOVQYWMFZTKMX-ONEGZZNKSA-N |
Synonym | 1-dimethylamino-2-nitroethylene, e-n,n-dimethyl-2-nitroethenamine, dimethyl e-2-nitroethenyl amine, n,n-dimethyl-2-nitroethenamine, 1-dimethylamino-2-nitroethene, dimethyl 2-nitroethenyl amine, n,n-dimethyl-2-nitroethylenamine, n,n-dimethyl-2-nitroethyleneamine, n,n-dimethyl-n-2-nitrovinyl amine |
PubChem CID | 637928 |
IUPAC Name | (E)-N,N-dimethyl-2-nitroethenamine |
SMILES | CN(C)C=C[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11424743
|
Thermo Scientific Alfa Aesar
A11637.03 |
1 g |
£23.10
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11434743
|
Thermo Scientific Alfa Aesar
A11637.06 |
5 g |
£83.20
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1190-92-7 | |
116.12 | |
JKOVQYWMFZTKMX-ONEGZZNKSA-N | |
637928 | |
CN(C)C=C[N+](=O)[O-] |
C4H8N2O2 | |
MFCD00051519 | |
1-dimethylamino-2-nitroethylene, e-n,n-dimethyl-2-nitroethenamine, dimethyl e-2-nitroethenyl amine, n,n-dimethyl-2-nitroethenamine, 1-dimethylamino-2-nitroethene, dimethyl 2-nitroethenyl amine, n,n-dimethyl-2-nitroethylenamine, n,n-dimethyl-2-nitroethyleneamine, n,n-dimethyl-n-2-nitrovinyl amine | |
(E)-N,N-dimethyl-2-nitroethenamine |
Specifications
1190-92-7 | |
C4H8N2O2 | |
1903136 | |
JKOVQYWMFZTKMX-ONEGZZNKSA-N | |
(E)-N,N-dimethyl-2-nitroethenamine | |
637928 | |
98% |
101°C to 105°C | |
MFCD00051519 | |
1-dimethylamino-2-nitroethylene, e-n,n-dimethyl-2-nitroethenamine, dimethyl e-2-nitroethenyl amine, n,n-dimethyl-2-nitroethenamine, 1-dimethylamino-2-nitroethene, dimethyl 2-nitroethenyl amine, n,n-dimethyl-2-nitroethylenamine, n,n-dimethyl-2-nitroethyleneamine, n,n-dimethyl-n-2-nitrovinyl amine | |
CN(C)C=C[N+](=O)[O-] | |
116.12 | |
116.12 | |
1-Dimethylamino-2-nitroethylene |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P272-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P333+P313-P362-P501c
H302-H315-H317-H318-H335
missing translation for 'einecsNumber' : 000-000-0
missing translation for 'tsca' : No
Recommended Storage : Keep cold
RUO – Research Use Only