Learn More
1-Dodecanethiol, 98%, Thermo Scientific Chemicals
£17.30 - £35.00
Chemical Identifiers
CAS | 112-55-0 |
---|---|
Molecular Formula | C12H26S |
Molecular Weight (g/mol) | 202.40 |
MDL Number | MFCD00004885 |
InChI Key | WNAHIZMDSQCWRP-UHFFFAOYSA-N |
Synonym | 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan |
PubChem CID | 8195 |
IUPAC Name | dodecane-1-thiol |
SMILES | CCCCCCCCCCCCS |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11440703
|
Thermo Scientific Alfa Aesar
A12741.22 |
100 g |
£17.30
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11450703
|
Thermo Scientific Alfa Aesar
A12741.36 |
500 g |
£35.00
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications1-Dodecanethiol is used in the preparation of hydrophobic or mixed self-assembled monolayers. It is also employed as a chain transfer agent for radical polymerization. Further, it is utilized as a polymerization inhibitor in polyurethane and neoprene adhesives, which finds application in the footwear industry. In addition to this, it acts as a protein regenerating agent used for the regeneration of native proteins from mercuribenzoate.
Solubility
Immiscible with water.
Notes
Air and moisture sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with bases, oxidizing agents, reducing agents and alkali metals.
Chemical Identifiers
112-55-0 | |
202.40 | |
WNAHIZMDSQCWRP-UHFFFAOYSA-N | |
8195 | |
CCCCCCCCCCCCS |
C12H26S | |
MFCD00004885 | |
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan | |
dodecane-1-thiol |
Specifications
112-55-0 | |
0.845 | |
133°C (271°F) | |
C12H26S | |
1.4601 | |
969337 | |
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan | |
WNAHIZMDSQCWRP-UHFFFAOYSA-N | |
dodecane-1-thiol | |
8195 | |
98% | |
1-Dodecanethiol |
°C to 7°C | |
276°C to 278°C | |
98% | |
CH3(CH2)11SH | |
MFCD00004885 | |
Air Sensitive | |
Immiscible with water. | |
CCCCCCCCCCCCS | |
202.40 | |
202.4 | |
Liquid |
Safety and Handling
GHS H Statement
H315-H319-H317-H335
P260-P264b-P271-P272-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P333+P313-P363-P501c
H314-H317-H335
missing translation for 'dotInformation' : Hazard Class: 8; Packaging Group: III
missing translation for 'einecsNumber' : 203-984-1
missing translation for 'rtecsNumber' : JR3155000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only