missing translation for 'onlineSavingsMsg'
Learn More

1-Dodecanethiol, 98%, Thermo Scientific Chemicals

£39.09 - £266.33

Chemical Identifiers

CAS 112-55-0
Molecular Formula C12H26S
Molecular Weight (g/mol) 202.40
MDL Number MFCD00004885
InChI Key WNAHIZMDSQCWRP-UHFFFAOYSA-N
Synonym 1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
PubChem CID 8195
IUPAC Name dodecane-1-thiol
SMILES CCCCCCCCCCCCS
View More Specs

Products 3
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10273950
View Documents Promotion Details
Thermo Scientific Acros
117625000
500 mL Glass bottle
£39.09
500mL
Please to purchase this item. Need a web account? Register with us today!
10358743
View Documents
Thermo Scientific Acros
117620025
2.5 L Glass bottle
£92.46
2.50 L
Please to purchase this item. Need a web account? Register with us today!
11965721
View Documents
Thermo Scientific Acros
117620100
10 L Plastic drum
£266.33
10 L
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

112-55-0
202.40
WNAHIZMDSQCWRP-UHFFFAOYSA-N
8195
CCCCCCCCCCCCS
C12H26S
MFCD00004885
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
dodecane-1-thiol

Specifications

112-55-0
100.0
0.8400g/mL
128°C
97.5% min. (GC)
CH3(CH2)11SH
MFCD00004885
0.84
Solubility in water: insoluble. Other solubilities: soluble in hydrocarbons, slightly soluble in alcohol
CCCCCCCCCCCCS
202.40
202.4
Liquid
97.5
-7.0°C
266.0°C to 283.0°C
Authentic
C12H26S
1.4580 to 1.4600
16,356
1-dodecanethiol, dodecyl mercaptan, dodecanethiol, n-dodecanethiol, lauryl mercaptan, n-dodecyl mercaptan, n-dodecylmercaptan, 1-mercaptododecane, n-lauryl mercaptan, 1-dodecyl mercaptan
WNAHIZMDSQCWRP-UHFFFAOYSA-N
dodecane-1-thiol
8195
98%
1-Dodecanethiol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes severe skin burns and eye damage.
May cause an allergic skin reaction.
Very toxic to aquatic life with long lasting effects.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with wat

GHS Signal Word: Danger

missing translation for 'einecsNumber' : 203-984-1

Product Identifier
  • 1-Dodecanethiol

Signal Word
  • Danger

Hazard Category
  • Skin corrosion/irritation Category 1 C
  • Serious eye damage/eye irritation Category 1
  • Skin sensitiser Category 1
  • LONG-TERM AQUATIC HAZARD Chronic 1

Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H410-Very toxic to aquatic life with long lasting effects.

Precautionary Statement
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only